Skip to Content
Merck

1.01596

Chemizorb® OH⁻

Synonym(s):

Absorbent for alkalis; Absorbent for bases

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.21
UNSPSC Code:
47131905
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder (or fine grains)

bulk density

30‑70 g/100 mL

storage temp.

no temp limit

General description

Absorbent and neutralizer for spilled alkalis, with indicator

Analysis Note

Identity: passes test
Bulk density: 30 - 70 g/100 ml
capacity (for caustic solution): ≥ 75 %

Legal Information

CHEMIZORB is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

13 - Non Combustible Solids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library