Skip to Content
Merck
All Photos(1)

Key Documents

07-024

Sigma-Aldrich

Anti-gp91-phox Antibody

Upstate®, from rabbit

Synonym(s):

Cytochrome b(558) subunit beta, Cytochrome b558 subunit beta, Heme-binding membrane glycoprotein gp91phox, NADPH oxidase, Neutrophil cytochrome b 91 kDa polypeptide, Superoxide-generating NADPH oxidase heavy chain subunit, chronic granulomatous disease,

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CYBB(1536)

General description

Cytochrome b-245, beta polypeptide (chronic granulomatous disease), also known as CYBB and P91-PHOX, is a human gene encoding a glycoprotein.
Cytochrome b (-245) is composed of cytochrome b alpha (CYBA) and beta (CYBB) chain. It has been proposed as a primary component of the microbicidal oxidase system of phagocytes. CYBB deficiency is one of five described biochemical defects associated with chronic granulomatous disease (CGD). In this disorder, there is decreased activity of phagocyte NADPH oxidase; neutrophils are able to phagocytize bacteria but cannot kill them in the phagocytic vacuoles.

Specificity

Detects gp91-phox, Mr 75 kDa. Depending on the type of lysate, the amount loaded, the gel and the blotting conditions, gp91-phox may run as a sharp band, Mr 75 kDa, or appear as a smear between 75-91 kDa.
Predicted to cross react with mouse.

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 548-560 of human gp91-phox (KQSISNSESGPRG). The immunizing sequence is identical in mouse, and it shares 11/13 sequence homology in bovine and 10/13 in porcine.

Application

Anti-gp91-phox Antibody is an antibody against gp91-phox for use in WB.
Immunoprecipitation:
Not recommended.
Research Category
Neuroscience
Research Sub Category
Oxidative Stress

Quality

Routinely evaluated by Western Blot on human neutrophil membranes.

Western Blot Analysis:
1-4 µg/mL of this lot detected gp91-phox in a HL60 membrane and RIPA cell lysate. A previous lot detected gp91-phox in human neutrophil membranes.

Target description

Varies. Depending on the type of lysate, the amount loaded, the gel and the blotting conditions, gp91-phox may run as a sharp band, Mr ~75 kDa, or appear as a smear between 75-91 kDa

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal IgG in buffer containing 0.07 M Tris-glycine, pH 7.4, 0.105 M NaCl, 0.035% sodium azide and 30% glycerol. Liquid at -20°C.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
For maximum recovery of product, centrifuge the vial prior to removing the cap.
Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. Note: Variabillity in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Analysis Note

Control
Lung tissue, human neutrophil membrane, HL60 membrane and RIPA cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Yu Lu et al.
Biochimica et biophysica acta, 1853(5), 965-974 (2015-01-21)
Our recent studies indicate that hydrogen peroxide (H2O2) only at high concentrations can cause oxidative stress in renal epithelial cells and induce apoptosis of podocytes. Consistently, the present study shows that H2O2, even at 1 mM, failed to induce intracellular
Stress reaction in outer segments of photoreceptors after blue light irradiation.
Roehlecke, C; Schumann, U; Ader, M; Brunssen, C; Bramke, S; Morawietz, H; Funk, RH
Testing null
Angiotensin Receptor Mediated Oxidative Stress Is Associated with Impaired Cardiac Redox Signaling and Mitochondrial Function in Insulin Resistant Rats.
Vazquez-Medina, Jose Pablo, et al.
American Journal of Physiology. Heart and Circulatory Physiology (2013)
Vascular aging in the longest-living rodent, the naked mole rat.
Csiszar, A; Labinskyy, N; Orosz, Z; Xiangmin, Z; Buffenstein, R; Ungvari, Z
American Journal of Physiology. Heart and Circulatory Physiology null
Interactions between the components of the human NADPH oxidase: intrigues in the phox family
Leusen, J H, et al
The Journal of Laboratory and Clinical Medicine, 128, 461-476 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service