Skip to Content
Merck
All Photos(1)

Key Documents

06-1340

Sigma-Aldrich

Anti-phospho-IKK-epsilon (Ser172) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Inhibitor of nuclear factor kappa-B kinase subunit epsilon, I-kappa-B kinase epsilon, IKK-E, IKK-epsilon, IkBKE, Inducible I kappa-B kinase, IKK-i

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
human, mouse
technique(s):
western blot: suitable
citations:
7

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

species reactivity (predicted by homology)

rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer172)

Gene Information

human ... IKBKE(9641)

General description

Inhibitor of nuclear factor kappa-B kinase subunit epsilon (UniProt: Q14164; also known as EC: 2.7.11.10, I-kappa-B kinase epsilon, IKK-E, IKK-epsilon, IkBKE, Inducible I kappa-B kinase, IKK-i) is encoded by the IKBKE (also known as IKKE, IKKI, KIAA0151) gene (Gene ID: 9641) in human. IKK-epsilon is a homodimeric serine/threonine kinase that is highly expressed in spleen, thymus, peripheral blood leukocytes, pancreas, and placenta. Its activity is induced by lipopolysaccharide and TNF-alpha. It plays an essential role in regulating inflammatory responses to viral infection, through the activation of the type I IFN, NF-kappa-B, and STAT signaling. IKK-epsilon can be either be auto-phosphorylated or phosphorylated by IKBKB/IKKB and phosphorylation at serine172 is enhanced by the interaction with ATP-dependent RNA helicase DDX3X. Its ATP-binding site resides in amino acids 15 to 23. IKK-epsilon is reported to phosphorylate inhibitors of NF-kappa-B leading to their ubiquitination and degradation. Following activation of viral RNA sensors, such as RIG-I-like receptors, it associates with DDX3X and phosphorylates interferon regulatory factors (IRFs), IRF3 and IRF7, as well as DDX3X. This activity allows subsequent homodimerization and nuclear translocation of the IRF3 leading to transcriptional activation of pro-inflammatory and antiviral genes including IFNB. In order to establish such an antiviral state, IKBKE forms several different complexes whose composition depends on the type of cell and cellular stimuli. Two isoforms of IKK-epsilon have been described that are produced by alternative splicing.

Specificity

This antibody recognizes IKK-epsilon phosphorylated at Ser172.

Immunogen

Epitope: Phosphorylated Ser172
Linear peptide corresponding to human IKKepsilon phosphorylated at Ser172.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-phospho-IKK-epsilon (Ser172) Antibody is validated for use in WB for the detection of phospho-IKK-epsilon (Ser172).

Quality

Evaluated by Western Blot in CD3+CD28 treated and untreated Jurkat cell lysates.

Western Blot Analysis: 1:1,000 dilution of this antibody detected IKK-epsilon on 10 µg of CD3+CD28 treated and untreated Jurkat cell lysates.

Target description

~90 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide and 30% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Analysis Note

Control
CD3+CD28 treated and untreated Jurkat cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measles virus suppresses RIG-I-like receptor activation in dendritic cells via DC-SIGN-mediated inhibition of PP1 phosphatases.
Mesman, AW; Zijlstra-Willems, EM; Kaptein, TM; de Swart, RL; Davis, ME; Ludlow, M; Duprex et al.
Cell host & microbe null
Sonja I Gringhuis et al.
Nature communications, 5, 3898-3898 (2014-05-29)
Carbohydrate-specific signalling through DC-SIGN provides dendritic cells with plasticity to tailor immunity to the nature of invading microbes. Here we demonstrate that recognition of fucose-expressing extracellular pathogens like Schistosoma mansoni and Helicobacter pylori by DC-SIGN favors T helper cell type-2
Yingjie Liu et al.
International journal of biological sciences, 17(5), 1353-1360 (2021-04-20)
Previous studies have described that NF-κB signaling mediated by NFκB-inducing kinase (NIK) plays a critical role of the differentiation of osteoclasts. We aim to explore the role of IKKe in methylprednisolone -induced osteonecrosis of the femoral head (ONFH). Methylprednisolone-induced ONFH
Gina Córdoba-David et al.
Frontiers in pharmacology, 13, 987979-987979 (2022-11-18)
The type I interferon (TI-IFN) pathway regulates innate immunity, inflammation, and apoptosis during infection. However, the contribution of the TI-IFN pathway or upstream signaling pathways to tubular injury in kidney disease is poorly understood. Upon observing evidence of activation of
Joris K Sprokholt et al.
PLoS pathogens, 13(11), e1006738-e1006738 (2017-12-01)
Follicular T helper cells (TFH) are fundamental in orchestrating effective antibody-mediated responses critical for immunity against viral infections and effective vaccines. However, it is unclear how virus infection leads to TFH induction. We here show that dengue virus (DENV) infection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service