Skip to Content
Merck

04-068

Anti-ATF1 Antibody, rabbit monoclonal

culture supernatant, Upstate®

Synonym(s):

Anti-EWS-ATF1, Anti-FUS/ATF-1, Anti-TREB36

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
monoclonal
Application:
WB
Citations:
7
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FUS(2521)

General description

37 kDa

Immunogen

KLH-conjugated synthetic peptide CPVTLTSQTTKTDDPQL

Application

Suggested dilution:
Western Blot: 1:1,000
Research Category
Signaling
Research Sub Category
MAP Kinases
This Anti-ATF1 Antibody, Rabbit is validated for use in WB for the detection of ATF1.

Biochem/physiol Actions

ATF1

Physical form

100 μL cell culture supernatant containing 0.05% Sodium Azide

Preparation Note

2 years at -20°C from date of shipment

Analysis Note

Routinely evaluated by immunoblot.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library