Skip to Content
Merck
All Photos(1)

Key Documents

H-095

Supelco

9-Hydroxyrisperidone-D4 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23D4H23FN4O3
CAS Number:
Molecular Weight:
430.51
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

OC(CCCN12)C1=NC(C)=C(C(C([2H])([2H])N3CCC(CC3)C4=NOC5=CC(F)=CC=C54)([2H])[2H])C2=O

InChI

1S/C23H27FN4O3/c1-14-17(23(30)28-9-2-3-19(29)22(28)25-14)8-12-27-10-6-15(7-11-27)21-18-5-4-16(24)13-20(18)31-26-21/h4-5,13,15,19,29H,2-3,6-12H2,1H3/i8D2,12D2

InChI key

PMXMIIMHBWHSKN-FJVJJXCISA-N

General description

An internal standard suitable for use in LC/MS or GC/MS applications including forensic analysis, clinical toxicology, urine drug testing, or isotope dilution methods. Also known as paliperidone, 9-Hydroxyrisperidone is sold under the brand name Invega, an atypical antipsychotic used for acute and maintenance treatment of schizophrenia. The drug is also the primary metabolite of risperidone, marketed as the atypical antipsychotic Risperdal®.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Risperdal is a registered trademark of Johnson & Johnson
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service