C-218S
Cannabinoid Mixture (Acids) - 6 Component solution
500 μg/mL (1% DIPEA and 0.05% Ascorbic acid in Acetonitrile, (each analyte)), certified reference material, ampule of 1 mL, Cerilliant®
About This Item
grade
certified reference material
Quality Level
feature
Snap-N-Spike®/Snap-N-Shoot®
composition
Cannabichromenic acid (CBCA), 500 μg/mL
Cannabidivarinic acid (CBDVA), 500 μg/mL
CBDA (Cannabidiolic acid), 500 μg/mL
CBGA (Cannabigerolic acid), 500 μg/mL
Tetrahydrocannabivarinic acid (THCVA), 500 μg/mL
THCA-A, 500 μg/mL
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
concentration
500 μg/mL (1% DIPEA and 0.05% Ascorbic acid in Acetonitrile, (each analyte))
format
multi-component solution
storage temp.
−70°C
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
35.6 °F - closed cup
Flash Point(C)
2.0 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service