Skip to Content
Merck
All Photos(1)

Key Documents

870721P

Avanti

20:4 Coenzyme A

Avanti Research - A Croda Brand 870721P, powder

Synonym(s):

(5Z,8Z,11Z,14Z-eicosatetraenoyl) Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H75N10O17P3S
CAS Number:
Molecular Weight:
1105.08
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (with stopper and crimp cap (870721P-5mg))

manufacturer/tradename

Avanti Research - A Croda Brand 870721P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCC/C=C\C/C=C\C/C=C\C/C=C\CCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C41H66N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-32(50)69-25-24-43-31(49)22-23-44-39(53)36(52)41(2,3)27-62-68(59,60)65-67(57,58)61-26-30-35(64-66(54,55)56)34(51)40(63-30)48-29-47-33-37(42)45-28-46-38(33)48;;;/h8-9,11-12,14-15,17-18,28-30,34-36,40,51-52H,4-7,10,13,16,19-27H2,1-3H3,(H,43,49)(H,44,53)(H,57,58)(H,59,60)(H2,42,45,46)(H2,54,55,56);3*1H3/b9-8-,12-11-,15-14-,18-17-;;;/t30-,34?,35+,36+,40-;;;/m1.../s1

InChI key

WINHAGOPPOAZSF-SCFPSPBGSA-N

Application

20:4 Coenzyme A has been used to determine the effects of lysophospholipid acyltransferase activities in adipocyte differentiation.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (870721P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lysophosphatidylcholine acyltransferase 3 is the key enzyme for incorporating arachidonic acid into glycerophospholipids during adipocyte differentiation
Eto M, et al.
International Journal of Molecular Sciences, 13(12), 16267-16280 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service