870294P
Avanti
18:0 PE Square
powder
Synonym(s):
1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-square (sodium salt)
About This Item
Recommended Products
Assay
99% (TLC)
form
powder
packaging
package of 1 × 10 mg (870294P-10MG)
package of 1 × 25 mg (870294P-25MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
shipped in
dry ice
storage temp.
−20°C
SMILES string
[H][C@@](COP([O-])(OCCNC1=C(OCC)C(C1=O)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[Na+]
InChI
1S/C47H86NO11P.Na/c1-4-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-42(49)56-39-41(40-58-60(53,54)57-38-37-48-44-45(51)46(52)47(44)55-6-3)59-43(50)36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-5-2;/h41,48H,4-40H2,1-3H3,(H,53,54);/q;+1/p-1/t41-;/m1./s1
InChI key
SYSGFMDVOBVKNR-QUEDAWNRSA-M
Packaging
5 mL Amber Glass Screw Cap Vial (870294P-25MG)
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service