Skip to Content
Merck
All Photos(1)

Key Documents

870012C

Avanti

18:1 MPB PE

Avanti Research - A Croda Brand 870012C

Synonym(s):

1,2-dioleoyl-sn-glycero-3-phosphoethanolamine-N-[4-(p-maleimidophenyl)butyramide] (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C55H88N2O11PNa
CAS Number:
Molecular Weight:
1007.26
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

liquid

packaging

pkg of 1 × 2.5 mL (870012C-25mg)
pkg of 2 × 4 mL (870012C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870012C

concentration

10 mg/mL (870012C-25mg)
25 mg/mL (870012C-200mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(CCCC(C=C1)=CC=C1N(C(C=C2)=O)C2=O)=O)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCC/C=C\CCCCCCCC)=O.[Na+]

InChI

1S/C55H89N2O11P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-36-54(61)65-46-50(68-55(62)37-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2)47-67-69(63,64)66-45-44-56-51(58)35-33-34-48-38-40-49(41-39-48)57-52(59)42-43-53(57)60;/h17-20,38-43,50H,3-16,21-37,44-47H2,1-2H3,(H,56,58)(H,63,64);/q;+1/p-1/b19-17-,20-18-;/t50-;/m1./s1

InChI key

IILKDCDPRGFBSN-VCRVZWGWSA-M

General description

Maleimidophenyl butyramide (MPB) phosphoethanolamine (PE), also known as DPPE-maleimide, contains maleimide, which is a thiol-reactive moiety and a good electrophile. MPB PE is implicated in liposome preparation.[1]

Application

18:1 MPB PE has been used in the preparation of nanoparticles.[2] It has also been used in the preparation of interbilayer-crosslinked multilamellar vesicles (ICMVs).[3]

Packaging

5 mL Clear Glass Sealed Ampule (870012C-200mg)
5 mL Clear Glass Sealed Ampule (870012C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel D-penicillamine carrying nanoparticles for metal chelation therapy in Alzheimer's and other CNS diseases
Cui Z, et al.
European Journal of Pharmaceutics and Biopharmaceutics, 59(2), 263-272 (2005)
The
Liposomes as tools in basic research and industry (1994)
Interbilayer-crosslinked multilamellar vesicles as synthetic vaccines for potent humoral and cellular immune responses
Moon J, et al.
Testing, 10(3), 243-243 (2011)
Emeka B Okeke et al.
Biomaterials, 238, 119836-119836 (2020-02-12)
Neutrophil elastase (NE) is a serine protease stored in the azurophilic granules of neutrophils and released into the extracellular milieu during inflammatory response or formation of neutrophil extracellular traps (NETs). Neutrophils release NETs to entrap pathogens by externalizing their cellular
Camilla Skyttner et al.
Langmuir : the ACS journal of surfaces and colloids, 34(22), 6529-6537 (2018-05-15)
Membrane-active peptides that enable the triggered release of liposomal cargo are of great interest for the development of liposome-based drug delivery systems but require peptide-lipid membrane interactions that are highly defined and tunable. To this end, we have explored the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service