Skip to Content
Merck
All Photos(1)

Key Documents

860696P

Avanti

Galactosyl(β) Sphingosine-d7

Avanti Research - A Croda Brand 860696P, powder

Synonym(s):

D-galactosyl-β-1,1’-D-erythro-sphingosine-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H40D7NO7
CAS Number:
Molecular Weight:
468.68
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.12

Assay

99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860696P-1MG)

manufacturer/tradename

Avanti Research - A Croda Brand 860696P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCC([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])(O)[C@](N)([H])CO[C@H](O1)[C@H](O)[C@@H](O)[C@H]([C@H]1CO)O

InChI

1S/C24H47NO7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-19(27)18(25)17-31-24-23(30)22(29)21(28)20(16-26)32-24/h14-15,18-24,26-30H,2-13,16-17,25H2,1H3/b15-14+/t18-,19+,20+,21-,22-,23+,24+/m0/s1/i1D3,2D2,3D2

InChI key

HHJTWTPUPVQKNA-RMWPRCMXSA-N

General description

Galactosyl(β) Sphingosine-d7 is a deuterated derivative of galactosyl(β) sphingosine. β- Galactosyl sphingosine, also known as psychosine, is localized in the brain.

Biochem/physiol Actions

β- Galactosyl Sphingosine is a toxic metabolite in Krabbe′s disease. It is hemolytic. β- Galactosyl Sphingosine also inhibits kinase C and phosphocholine cytidyltransferase (CTP).

Packaging

5 mL Amber Glass Screw Cap Vial

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of inhibition of purified and mitochondrial cytochrome c oxidase by psychosine (beta-galactosylsphingosine)
Cooper C E, et al.
The Biochemical Journal, 290(1), 139-144 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service