Skip to Content
Merck
All Photos(1)

Key Documents

860580P

Avanti

02:0 SM (d18:1/2:0)

Avanti Research - A Croda Brand 860580P, powder

Synonym(s):

N-acetyl-D-erythro-sphingosylphosphorylcholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H51N2O6P
CAS Number:
Molecular Weight:
506.66
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (860580P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860580P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(C)=O)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C25H51N2O6P/c1-6-7-8-9-10-11-12-13-14-15-16-17-18-19-25(29)24(26-23(2)28)22-33-34(30,31)32-21-20-27(3,4)5/h18-19,24-25,29H,6-17,20-22H2,1-5H3,(H-,26,28,30,31)/b19-18+/t24-,25+/m0/s1

InChI key

GTZSEZHHSPWFFN-WHUSYDGZSA-N

General description

Sphingomyelin is abundantly present in the cell membranes, particularly in myelin sheath and red blood cells. It has a ceramide unit with a phosphorylcholine moiety linked to position 1.

Application

02:0 SM (d18:1/2:0) is suitable for use in liposomes to measure ostreolysin A (OlyA) binding in order to infer the effects of phosphatidylcholines (PCs) on sphingomyelin (SM)/cholesterol interactions.

Biochem/physiol Actions

Sphingomyelin (SM) has high cholesterol affinity, which aids localization and interactions of proteins. It functions as a precursor to ceramides and other sphingolipid metabolites.

Packaging

5 mL Clear Glass Sealed Ampule (860580P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ceramide and neurodegeneration: susceptibility of neurons and oligodendrocytes to cell damage and death
Jana A, et al.
Journal of the Neurological Sciences, 278(1-2), 5-15 (2009)
Molecular discrimination between two conformations of sphingomyelin in plasma membranes
Endapally S, et al.
Cell, 176(5), 1040-1053 (2019)
Delphine Milhas et al.
FEBS letters, 584(9), 1887-1894 (2009-10-28)
The plasma membrane (PM) is a major resource for production of bioactive lipids and contains a large proportion of the cellular sphingomyelin (SM) content. Consequently, the regulation of SM levels at the PM by enzymes such as sphingomyelinase (SMase) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service