Skip to Content
Merck
All Photos(2)

Key Documents

840514C

Avanti

16:0-18:1 Phosphatidylethanol

Avanti Research - A Croda Brand 840514C

Synonym(s):

1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanol (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H74NaO8P
CAS Number:
Molecular Weight:
724.96
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840514C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840514C

concentration

10 mg/mL (840514C-25mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC)=O)(OC(CCCCCCC/C=C\CCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C39H75O8P.Na/c1-4-7-9-11-13-15-17-19-20-22-24-26-28-30-32-34-39(41)47-37(36-46-48(42,43)45-6-3)35-44-38(40)33-31-29-27-25-23-21-18-16-14-12-10-8-5-2;/h19-20,37H,4-18,21-36H2,1-3H3,(H,42,43);/q;+1/p-1/b20-19-;/t37-;/m1./s1

InChI key

JFUGSRIBTPUDOW-VYMFHCNISA-M

General description

Phosphatidylethanol (PEth) is a phospholipid, that is generated in the the presence of ethanol in cell membranes. PEths like PEth 16:0/18:1, PEth 18:1/18:1, and PEth 16:0/16:0 possesses a common phosphoethanol head. Two fatty acid chains of variable length and degree of saturation are bound to this head group. It is widely distributed in gram-positive organisms.
Phosphatidylethanol (PEth) represents a group of glycerophospholipid homologues generated by substitution of ethanol for the lipid headgroup by the phospholipase D enzyme. Since the formation of PEth is specifically dependent on ethanol, the diagnostic specificity of PEth as an alcohol biomarker is theoretically 100%.

Application

16:0-18:1 Phosphatidylethanol is suitable for use in the preparation of liposomes used for permeability studies.

Biochem/physiol Actions

Phosphatidylethanols (PEths) serves as crucial biomarkers of alcohol consumption.

Packaging

5 mL Clear Glass Sealed Ampule (840514C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Interactions of oritavancin, a new lipoglycopeptide derived from vancomycin, with phospholipid bilayers: effect on membrane permeability and nanoscale lipid membrane organization
Domenech O, et al.
Biochimica et Biophysica Acta - Biomembranes, 1788(9), 1832-1840 (2009)
David Wensbo Posaric et al.
Analytical and bioanalytical chemistry, 406(19), 4735-4744 (2014-05-29)
Various phosphatidylethanol (PEth) derivatives, the corresponding reversed positional isomers (RPI-PEths), lyso-PEth-16:0, and penta-deuterium-labeled PEth analogs (d5-PEths), were synthesized by enzyme-independent synthetic routes. A general solvent system consisting of a mixture of acetone-d6 and methanol-d4 (97:3; v/v) was found to provide
The detection of 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanol and ethyl glucuronide in human umbilical cord
Jones J, et al.
American Journal of Analytical Chemistry, 3(12), 800-800 (2012)
Alexandra Schröck et al.
Bioanalysis, 6(17), 2285-2294 (2014-11-11)
For early diagnosis and therapy of alcohol-related disorders, alcohol biomarkers are highly valuable. Concerning specificity, indirect markers can be influenced by nonethanol-related factors, whereas direct markers are only formed after ethanol consumption. Sensitivity of the direct markers depends on cut-offs
Anders Isaksson et al.
Drug testing and analysis, 3(4), 195-200 (2011-03-26)
Phosphatidylethanol (PEth) represents a group of glycerophospholipid homologues where ethanol by phospholipase D has been bound at the position that normally contains an amino-alcohol. Since the formation of PEth is specifically dependent on ethanol, the diagnostic specificity of PEth as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service