Skip to Content
Merck
All Photos(3)

Key Documents

840052P

Avanti

Heart PC

Avanti Research - A Croda Brand 840052P, powder

Synonym(s):

L-α-phosphatidylcholine (Heart, Bovine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 2 × 100 mg (840052P-200mg)
pkg of 1 × 25 mg (840052P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840052P

lipid type

phosphoglycerides
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OC(CCCCCCC/C=C\C/C=C\CCCCC)=O)COC(CCCCCCCCCCCCCCC)=O)=O

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

General description

Contains ~40% plasmalogen ether lipid
Phosphatidylcholine (PC) is a glycerophospholipid present abundantly in eukaryotes. It comprises 40-50% of the total phospholipid content in the cell membrane. PC is synthesized by the Kennedy pathway.

Application

Heart PC has been used in the preparation of phospholipid reference mixture.

Biochem/physiol Actions

Phosphatidylcholine (PC) in the liver is responsible for the production of very low density lipoproteins (VLDL). It is an important source for the formation of lipid mediators and secondary messengers.

Packaging

5 mL Clear Glass Sealed Ampule (840052P-200mg)
5 mL Clear Glass Sealed Ampule (840052P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phosphatidylethanolamine and phosphatidylcholine biosynthesis by the Kennedy pathway occurs at different sites in Trypanosoma brucei
Farine L, et al.
Scientific reports, 5(6), 16787-16787 (2015)
Expression of Shiga toxin 2e glycosphingolipid receptors of primary porcine brain endothelial cells and toxin-mediated breakdown of the blood-brain barrier
Meisen I, et al.
Glycobiology, 23(6), 745-759 (2013)
Marianne S Jurkowitz et al.
Biochimica et biophysica acta, 1848(2), 742-751 (2014-12-03)
Lysoplasmalogenase catalyzes hydrolytic cleavage of the vinyl-ether bond of lysoplasmalogen to yield fatty aldehyde and glycerophospho-ethanolamine or glycerophospho-choline. We recently purified lysoplasmalogenase from rat liver microsomes and identified the protein as TMEM86B, an integral membrane protein that is a member
The critical role of phosphatidylcholine and phosphatidylethanolamine metabolism in health and disease.
van der Veen J N, et al.
Biochimica et Biophysica Acta - Biomembranes, 1859(9 Pt B), 1558-1558 (2017)
Ivan U Kouzel et al.
Journal of lipid research, 54(3), 692-710 (2012-12-19)
Glycosphingolipids (GSLs) of the globo-series constitute specific receptors for Shiga toxins (Stxs) released by certain types of pathogenic Escherichia coli strains. Stx-loaded leukocytes may act as transporter cells in the blood and transfer the toxin to endothelial target cells. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service