Skip to Content
Merck
All Photos(3)

Key Documents

840052C

Avanti

Heart PC

Avanti Research - A Croda Brand 840052C

Synonym(s):

L-α-phosphatidylcholine (Heart, Bovine)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (840052C-25mg)
pkg of 2 × 4 mL (840052C-200mg)
pkg of 5 × 4 mL (840052C-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand 840052C

concentration

10 mg/mL (840052C-25mg)
25 mg/mL (840052C-200mg)
25 mg/mL (840052C-500mg)

lipid type

phosphoglycerides
phospholipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

General description

Contains ~40% plasmalogen ether lipid
Phosphatidylcholine (PC) is a phospholipid, that comprises 40-50% of the total phospholipid content in the cell membrane. It is an abundant phospholipid in all mammalian cells and cellular organelles. PC is a glycerophospholipid. It is synthesized by the Kennedy pathway.

Application

Heart PC has been used in the Mycobacterium lipid extraction and in β- Hexosaminidase release and calcium mobilization assays.

Biochem/physiol Actions

Phosphatidylcholine (PC) in the liver is responsible for the production of very low density lipoproteins (VLDL).

Packaging

5 mL Clear Glass Sealed Ampule (840052C-200mg)
5 mL Clear Glass Sealed Ampule (840052C-25mg)
5 mL Clear Glass Sealed Ampule (840052C-500mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Liver,Kidney, Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The critical role of phosphatidylcholine and phosphatidylethanolamine metabolism in health and disease.
van der Veen J N, et al.
Biochimica et Biophysica Acta - Biomembranes, 1859(9 Pt B), 1558-1558 (2017)
Marianne S Jurkowitz et al.
Biochimica et biophysica acta, 1848(2), 742-751 (2014-12-03)
Lysoplasmalogenase catalyzes hydrolytic cleavage of the vinyl-ether bond of lysoplasmalogen to yield fatty aldehyde and glycerophospho-ethanolamine or glycerophospho-choline. We recently purified lysoplasmalogenase from rat liver microsomes and identified the protein as TMEM86B, an integral membrane protein that is a member
Mycobacterial Lipids Induce Calcium Mobilization and Degranulation of Mast Cells
Torres-Atencio I, et al.
American Journal of Respiratory and Critical Care Medicine, 198(6), 813-816 (2018)
Phosphatidylethanolamine and phosphatidylcholine biosynthesis by the Kennedy pathway occurs at different sites in Trypanosoma brucei
Farine L, et al.
Scientific Reports, 5(9 Pt B), 16787-16787 (2015)
Ivan U Kouzel et al.
Journal of lipid research, 54(3), 692-710 (2012-12-19)
Glycosphingolipids (GSLs) of the globo-series constitute specific receptors for Shiga toxins (Stxs) released by certain types of pathogenic Escherichia coli strains. Stx-loaded leukocytes may act as transporter cells in the blood and transfer the toxin to endothelial target cells. Therefore

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service