Skip to Content
Merck
All Photos(2)

Key Documents

810155P

Avanti

18:1-06:0 NBD PE

Avanti Research - A Croda Brand

Synonym(s):

1-oleoyl-2-{6-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]hexanoyl}-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H58N5O11P
CAS Number:
Molecular Weight:
755.84
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810155P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) is a fluorescently labelled lipid with NBD attached to phosphatidylethanolamine. It is localized at the membrane interface. Phosphoethanolamine is the second most abundant, metabolically related aminophospholipid and is mostly enriched in brain.

Application

18:1-06:0 NBD PE may be used to analyse the temporal fluorescent pattern of cholesterol-E- Nitrobenzoxadiazole (Chol-E-NBD).

Biochem/physiol Actions

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) is used as a fluorescent analog in the studies of organization and dynamics of membranes. Phosphoethanolamine (PE) functions as a substrate for a number of phospholipids of the cell membrane.

Packaging

5 mL Amber Glass Screw Cap Vial (810155P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic phosphoethanolamine induces cell cycle arrest and apoptosis in human breast cancer MCF-7 cells through the mitochondrial pathway
Ferreira AK, et al.
Biomedicine & Preventive Nutrition, 67(6), 481-487 (2013)
Fluorescent methods to study biological membranes, 37-50 (2012)
Cardiolipin and phosphatidylethanolamine role in dibucaine interaction with the mitochondrial membrane
Lopes SC, et al.
Biochim. Biophys. Acta Gen. Subj., 1861(6), 1152-1161 (2019)
Synthesis and validation of novel cholesterol-based fluorescent lipids designed to observe the cellular trafficking of cationic liposomes
Kim BK, et al.
Bioorganic & Medicinal Chemistry, 25(18), 3893-3896 (2015)
Formation and function of phosphatidylserine and phosphatidylethanolamine in mammalian cells
Vance JE and Tasseva G
Biochim. Biophys. Acta Gen. Subj., 1831(3), 543-554 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service