Skip to Content
Merck
All Photos(2)

Key Documents

810152P

Avanti

14:0-12:0 NBD PE

Avanti Research - A Croda Brand 810152P, powder

Synonym(s):

1-myristoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H64N5O11P
CAS Number:
Molecular Weight:
785.90
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810152P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810152P

shipped in

dry ice

storage temp.

−20°C

General description

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) is a fluorescently labeled lipid localized to the membrane interface. The molecule contains NBD as a fluorophore moiety linked to the headgroup of phosphatidylethanolamine. Phosphoethanolamine is the second most abundant, metabolically related aminophospholipid. It is mostly enriched in brain.

Biochem/physiol Actions

Phosphoethanolamine present in cardiac myocytes plays a vital role in the prevention of cell damage after ischemia. N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) supports the sensing of the Red edge excitation shift (REES) phenomenon with the unique motional and dielectric properties it possesses.

Packaging

5 mL Amber Glass Screw Cap Vial (810152P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of NBD-labeled lipids in membrane and cell biology
Fluorescent methods to study biological membranes, 37-50 (2012)
S C Lopes et al.
Biochimica et biophysica acta. Biomembranes, 1861(6), 1152-1161 (2019-03-07)
Mitochondrial membranes are pointed out as the site of cardiotoxic action of local anaesthetics. Its three main phospholipids components are phosphatidylcholine, phosphatidylethanolamine and cardiolipin. Cardiolipins, in eukaryotes, are only found in mitochondria and are essential for the maintenance of its
Jean E Vance et al.
Biochimica et biophysica acta, 1831(3), 543-554 (2012-09-11)
Phosphatidylserine (PS) and phosphatidylethanolamine (PE) are metabolically related membrane aminophospholipids. In mammalian cells, PS is required for targeting and function of several intracellular signaling proteins. Moreover, PS is asymmetrically distributed in the plasma membrane. Although PS is highly enriched in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service