Skip to Content
Merck
All Photos(2)

Key Documents

810128P

Avanti

12:0 Lyso NBD PC

Avanti Research - A Croda Brand 810128P, powder

Synonym(s):

1-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-2-hydroxy-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44N5O10P
CAS Number:
Molecular Weight:
617.63
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (LPC; may contain up to 10% of the 2-LPC isomer, TLC)

form

powder

packaging

pkg of 1 × 1 mg (810128P-1mg)
pkg of 5 × 1 mg (810128P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810128P

shipped in

dry ice

storage temp.

−20°C

Application

12:0 Lyso NBD PC has been used in the NBD-LPC (7-nitrobenz-2-oxa-1,3-diazol-4-yl - Lysophosphatidylcholine) uptake assays where the fluorescence intensity of the NBD-LPC signal can be measured. It may also be used as a substrate to measure the enzymatic activity of recombinant human lysophosphatidylcholine acyltransferase 3 (rhLPCAT3).

Packaging

5 mL Amber Screw Cap Bottle (810128P-1mg)
5 mL Amber Screw Cap Bottle (810128P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shweta Tiwary et al.
Scientific reports, 8(1), 8267-8267 (2018-05-31)
Disruption of the blood-brain barrier (BBB) by cancer cells is linked to metastatic tumor initiation and progression; however, the pathways that drive these events remain poorly understood. Here, we have developed novel patient-derived xenograft (PDX) models of brain metastases that
Lauren P Saunders et al.
The Journal of biological chemistry, 286(34), 30130-30141 (2011-07-02)
Autotaxin (ATX) is a secreted lysophospholipase D that hydrolyzes lysophosphatidylcholine (LPC) into lysophosphatidic acid (LPA), initiating signaling cascades leading to cancer metastasis, wound healing, and angiogenesis. Knowledge of the pathway and kinetics of LPA synthesis by ATX is critical for
Xinming Du et al.
FEBS open bio, 9(10), 1734-1743 (2019-08-04)
Lysophosphatidylcholine acyltransferase 3 (LPCAT3) is an important enzyme in phospholipid remodeling, a process that influences the biophysical properties of cell membranes and thus cell function. Multiple lines of evidence suggest that LPCAT3 is involved in several diseases, including atherosclerosis, non-alcoholic
Stephen M Baumler et al.
Langmuir : the ACS journal of surfaces and colloids, 33(12), 2986-2992 (2017-03-08)
The translational diffusion dynamics of the zwitterionic lipid 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) at a planar phosphorylated support surface containing metal ions (Mg2+, Ca2+, Ba2+, Ni2+, Zn2+, Cd2+, Zr4+) was investigated using X-ray photoelectron spectroscopy (XPS) and fluorescence recovery after photobleaching (FRAP). Fluorescence

Articles

The critical micelle concentration (CMC) can be approximately defined as the lipid monomer concentration at which appreciable amounts (>5% of total) of micellar aggregates first begin to appear in the equilibrium: nM1<=>Mn

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service