810104P
Avanti
palmitic acid - lissamine rhodamine
Avanti Research™ - A Croda Brand 810104P, powder
Synonym(s):
[16-N-(lissamine rhodamine β sulfonyl)amino] palmitic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C43H61N3O8S2
CAS Number:
Molecular Weight:
812.09
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Recommended Products
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 1 mg (810104P-1mg)
manufacturer/tradename
Avanti Research™ - A Croda Brand 810104P
shipped in
dry ice
storage temp.
−20°C
General description
Palmitic acid (PA), a saturated fatty acid, seen in human body accounts for 20−30% of total fatty acids. It can be supplied through diet or produced endogenously from other fatty acids, carbohydrates and amino acids. PA is an important constituent of palm oil. Lissamine rhodamine is a red-emitting fluorophore, mainly used in immunofluorescence studies.
Packaging
5 mL Amber Glass Screw Cap Vial (810104P-1mg)
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Palmitic acid: Physiological role, metabolism and nutritional implications
Carta G, et al.
The Journal of Physiology (2017)
Which fluorophore is brightest? A comparison of the staining obtained using fluorescein, tetramethylrhodamine, lissamine rhodamine, Texas red, and cyanine 3.18
Wessendorf MW and Brelje TC
Histochemistry (1992)
Palmitic acid: Physiological role, metabolism and nutritional implications
Carta G, et al.
Frontiers in Physiology, 8, 902-902 (2017)
Which fluorophore is brightest? A comparison of the staining obtained using fluorescein, tetramethylrhodamine, lissamine rhodamine, Texas red, and cyanine 3.18
Wessendorf MW and Brelje TC
Histochemistry, 98(2) (1992)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service