Z742016
Duran® Youtility Screw CapGL 45
cyan, polypropylene (food grade)
Synonym(s):
Duran® Youtility
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Recommended Products
material
polypropylene (food grade)
packaging
pkg of 10 ea
manufacturer/tradename
DURAN® 292292802
color
, cyan
Looking for similar products? Visit Product Comparison Guide
General description
- Specially shaped gripping zones on both sides of the bottle for easier and safer handling
- Opening or closing of the bottle is faster, and fully compatible with DIN GL 45 closures and other accessories
- Slimmer YOUTILITY bottle shape allows a more optimal use of limited space in autoclaves and laboratory refrigerators
- Pre-defined labelling area is compatible with DURAN self-adhesive YOUTILITY bottle labels
- Nominal volume is shown at the top of the easy-to-read graduation scale for fast determination of the volumes
- Sequentially sized bottles are available in four volumes
- Moulded from the DURAN borosilicate 3.3 pharmacopeia type 1 neutral glass
- Retrace Code for full product traceability
- Each bottle is supplied as a complete system, with a pouring ring (cyan polypropylene) and a GL 45 cap (cyan polypropylene)
Features and Benefits
- Faster opening and closing thanks to the shortened thread, which is still compatible with all GL 45 laboratory bottles
- The optimized, ergonomic shape of the screw cap has comfortable grooves and ridges for a more efficient and easier tightening or removal, especially with smaller sized or gloved hands
- The tried and tested cap plug sealing with sealing lip system ensures a liquid tight seal
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service