Key Documents
Z682330
Ace rotary evaporator condenser complete "A" assembly
glass, fits Büchi® models R-200/R-205 and R-114/R-144
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
material
glass
manufacturer/tradename
Büchi equivalent, 40633
Related Categories
General description
The complete "A" evaporator condenser assembly includes the main glass condenser body with inner coil for angled installation, GL threaded caps and hose barbs. Bottom joint is 35/20. It is designed to fit Büchi® evaporator models 200/205 and series 114-144.
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)