Key Documents
Z657220
IKA® overhead stirrer accessories
R271 boss head clamp
About This Item
UNSPSC Code:
41122800
NACRES:
NB.72
packaging
pack of 1 ea
manufacturer/tradename
IKA 0002664000
General description
The boss head is used for the safe fixing of drives to the stands. It resists strongest mechanical demands.
Stand diam. 34 mm, Bracket diam. 16mm
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)