Z303887
Quickfit® flat flange lids for reaction flasks
center joint: ST/NS 29/32
Synonym(s):
DA4/100
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Recommended Products
material
borosilicate glass
Joint
center joint: ST/NS29/32
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Quickfit DA 4/100
availability
not available in USA and Canada
flange bore
100 mm
General description
Single and multi-socket lids for reaction flasks for simple extraction, reflux or for the introduction of a variety of stirrers, dropping funnels, temperature probes etc.
Use clip to secure lid to flasks, purchase separately.
Use clip to secure lid to flasks, purchase separately.
Features and Benefits
- Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)
- With precision ground flat-flange joints to fit Quickfit wide neck reaction flasks and culture vessels
- Fits Quickfit wide neck reaction flasks, culture vessels and large capacity Soxhlet extractors
- Each lid with one or more ground socket joints to accept other ground joint components
- Lids should be secured to flasks with relevant metal clamps, JC75F or JC100F, depending on flange diameter
- Please note that these lids are not compatible with the DURAN® range of flat flange products
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)