Z265233
TLC saturation pads
size 10 cm × 20 cm
Synonym(s):
TLC developing chamber saturation pads
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
Recommended Products
packaging
pkg of 100 ea
manufacturer/tradename
Analtech 8112
technique(s)
thin layer chromatography (TLC): suitable
size
10 cm × 20 cm
Looking for similar products? Visit Product Comparison Guide
General description
Placement of pads with solvent in the development chamber helps to assure rapid and uniform saturation of the atmosphere with solvent vapor. Pads improve reproducibility and reduce "edge effects".
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service