W357804
2-Methyl-4-propyl-1,3-oxathiane, mixture of cis and trans
≥98%, FG
Synonym(s):
2-tropical oxathiane, Tropathiane
About This Item
Fragrance grade
Halal
Kosher
Recommended Products
biological source
synthetic
Quality Level
grade
FG
Fragrance grade
Halal
Kosher
Agency
follows IFRA guidelines
meets purity specifications of JECFA
reg. compliance
EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
Assay
≥98%
refractive index
n20/D 1.4800 (lit.)
bp
218-220 °C (lit.)
density
0.975 g/mL at 25 °C (lit.)
application(s)
flavors and fragrances
Documentation
see Safety & Documentation for available documents
food allergen
no known allergens
fragrance allergen
no known allergens
Organoleptic
green; tropical
SMILES string
CCCC1CCOC(C)S1
InChI
1S/C8H16OS/c1-3-4-8-5-6-9-7(2)10-8/h7-8H,3-6H2,1-2H3
InChI key
GKGOLPMYJJXRGD-UHFFFAOYSA-N
General description
Application
- Evolution and Correlation of cis-2-Methyl-4-propyl-1,3-oxathiane, Varietal Thiols, and Acetaldehyde during Fermentation of Sauvignon blanc Juice.: This research highlights the development and interaction of cis-2-Methyl-4-propyl-1,3-oxathiane with varietal thiols and acetaldehyde in Sauvignon blanc during fermentation, emphasizing its impact on the wine′s aroma profile (Wang et al., 2020).
Signal Word
Warning
Hazard Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
122.0 °F - closed cup
Flash Point(C)
50 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service