Skip to Content
Merck
All Photos(1)

Key Documents

W327107

Sigma-Aldrich

2,3-Dimethylpyrazine

≥95%, FCC, FG

Synonym(s):

2,3-dimethyl-1,4-diazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2
CAS Number:
Molecular Weight:
108.14
FEMA Number:
3271
EC Number:
Council of Europe no.:
11323
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.050
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥95%

refractive index

n20/D 1.507 (lit.)

bp

156 °C (lit.)

density

1.011 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

coffee; nutty; roasted

SMILES string

Cc1nccnc1C

InChI

1S/C6H8N2/c1-5-6(2)8-4-3-7-5/h3-4H,1-2H3

InChI key

OXQOBQJCDNLAPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Characterization of key aroma compounds in Chinese smoked duck by SAFE-GC-O-MS and aroma-recombination experiments.: This study identifies and quantifies the key volatile compounds contributing to the aroma profile of Chinese smoked duck, utilizing sophisticated analytical techniques like SAFE-GC-O-MS, highlighting the impact of 2,3-Dimethylpyrazine among other compounds (Liu et al., 2023).

  • Exploring the formation and retention of aroma compounds in ready-to-eat roasted pork from four thermal methods: A lipidomics and heat transfer analysis.: This research explores how different cooking methods affect the aroma profiles of roasted pork, including the formation of 2,3-Dimethylpyrazine, providing insights into optimizing flavor through lipidomics and heat transfer analyses (Liu et al., 2024).

  • Flavor evolution of normal- and low-fat Chinese sausage during natural fermentation.: This article investigates the dynamic changes in flavor compounds, including 2,3-Dimethylpyrazine, during the fermentation of Chinese sausages, offering valuable information on flavor development in fermented foods (Xing et al., 2023).

  • Aroma generation in sponge cakes: The influence of sucrose particle size and sucrose source.: This study examines how different types of sucrose affect the generation of key aroma compounds such as 2,3-Dimethylpyrazine in sponge cakes, shedding light on ingredient impacts on bakery product flavors (Garvey et al., 2023).

  • Application of Maillard Reaction Products Derived Only from Enzymatically Hydrolyzed Sesame Meal to Enhance the Flavor and Oxidative Stability of Sesame Oil.: This research discusses the use of Maillard reaction products, including 2,3-Dimethylpyrazine, from sesame meal to enhance both the flavor profile and stability of sesame oil, illustrating the potential of enzymatic processes in flavor enhancement (Ma et al., 2022).


Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Issei Yokoyama et al.
Scientific reports, 10(1), 13931-13931 (2020-08-20)
The effects of odors generated by the Maillard reaction from amino acids and reducing sugars on physiological parameters (blood pressure, heart rate, and oxidative stress levels) in Wistar rats were investigated in the present study. The Maillard reaction samples were
Joachim Morrens et al.
Neuron, 106(1), 142-153 (2020-02-07)
Dopamine neurons mediate the association of conditioned stimuli (CS) with reward (unconditioned stimuli, US) by signaling the discrepancy between predicted and actual reward during the US. Some theoretical models suggest that learning is also influenced by the salience or associability
Nobuo Ochiai et al.
Journal of chromatography. A, 1421, 103-113 (2015-08-16)
An extension of multi-volatile method (MVM) technology using the combination of a standard dynamic headspace (DHS) configuration, and a modified DHS configuration incorporating an additional vacuum module, was developed for milliliter injection volume of aqueous sample with full sample evaporation.
Gustavo Luis Leonardo Scalone et al.
Journal of agricultural and food chemistry, 63(22), 5364-5372 (2015-05-15)
Pyrazines are specific Maillard reaction compounds known to contribute to the unique aroma of many products. Most studies concerning the generation of pyrazines in the Maillard reaction have focused on amino acids, while little information is available on the impact

Protocols

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Separation of various pyrazines and sulfur compounds including carbon disulfide and dimethyl disulfide.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service