Skip to Content
Merck
All Photos(1)

Key Documents

W274518

Sigma-Aldrich

Methyl salicylate

≥98%, FCC, FG

Synonym(s):

2-Hydroxybenzoic acid methyl ester, Methyl 2-hydroxybenzoate, Oil of wintergreen, Wintergreen oil

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2CH3
CAS Number:
Molecular Weight:
152.15
FEMA Number:
2745
Beilstein:
971516
EC Number:
Council of Europe no.:
433
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
9.749
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC
FDA 21 CFR 117
FDA 21 CFR 175.105

vapor density

5.26 (vs air)

vapor pressure

1 mmHg ( 54 °C)

Assay

≥98%

form

liquid

autoignition temp.

847 °F

refractive index

n20/D 1.536 (lit.)

bp

222 °C (lit.)

mp

−8-−7 °C (lit.)

density

1.174 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

spicy; minty; cooling; sweet; wintergreen

SMILES string

COC(=O)c1ccccc1O

InChI

1S/C8H8O3/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5,9H,1H3

InChI key

OSWPMRLSEDHDFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl salicylate is one of the main volatile flavor components in tomatoes, sapodilla and unburned tobacco.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Volatile flavor components of sapodilla fruit (Achras sapota L).
MacLeod AJ & Gonzales de Troconis N.
Journal of Agricultural and Food Chemistry, 30(3), 515-517 (1982)
Matthes Kenning et al.
Zoological letters, 5, 3-3 (2019-01-19)
In the context of evolutionary arthopodial transformations, centipede ultimate legs exhibit a plethora of morphological modifications and behavioral adaptations. Many species possess significantly elongated, thickened, or pincer-like ultimate legs. They are frequently sexually dimorphic, indicating a role in courtship and
Quantitation of ten flavor compounds in unburned tobacco products.
Lisko JG, et al.
Analytical Methods : Advancing Methods and Applications, 6(13), 4698-4704 (2014)
Flavor chemistry of tomato volatiles.
Kazeniac SJ & Hall RM
Journal of Food Science, 35(5), 519-530 (1970)
Hsu Shao Feng et al.
Journal of clinical periodontology, 38(7), 637-643 (2011-04-01)
To evaluate the clinical efficacy of subgingival ultrasonic instrumentation irrigated with essential oils (EOs) of residual periodontal pockets. Sixty-four individuals with chronic periodontitis were invited to participate in this randomized, double-blind, parallel, and placebo-controlled clinical trial. All subjects received non-surgical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service