Skip to Content
Merck
All Photos(1)

Key Documents

T58203

Sigma-Aldrich

2,4,6-Tris(dimethylaminomethyl)phenol

95%

Synonym(s):

DMP-30

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2NCH2]3C6H2OH
CAS Number:
Molecular Weight:
265.39
Beilstein:
795751
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

vapor pressure

<0.01 mmHg ( 21 °C)

Assay

95%

refractive index

n20/D 1.516 (lit.)

bp

130-135 °C/1 mmHg (lit.)

density

0.969 g/mL at 25 °C (lit.)

SMILES string

CN(C)Cc1cc(CN(C)C)c(O)c(CN(C)C)c1

InChI

1S/C15H27N3O/c1-16(2)9-12-7-13(10-17(3)4)15(19)14(8-12)11-18(5)6/h7-8,19H,9-11H2,1-6H3

InChI key

AHDSRXYHVZECER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,4,6-Tris(dimethylaminomethyl)phenol can be used:
  • In the synthesis of water-soluble metal phthalocyanines bearing twelve dimethylamino groups.
  • To prepare phenolate anion-based branched/cross-linked anion exchange membranes (AEMs).
  • As a curing agent in the production of epoxy membranes.

Legal Information

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

300.2 °F - closed cup

Flash Point(C)

149 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Water soluble novel phthalocyanines containing dodeca-amino groups.
Sesalan B S, et al.
Dyes and Pigments, 79(3), 259-264 (2008)
Phenolate anion-based branched/cross-linked poly (arylene ether sulfone) hydroxide exchange membranes.
Zhou J, et al.
International Journal of Hydrogen Energy, 41(13), 5765-5775 (2016)
Permeability and transport model of epoxy membranes with various surface morphologies.
Luo Y S, et al.
Journal of Applied Polymer Science, 130(4), 2938-2946 (2013)
S Bratosin-Guttman
Journal of structural biology, 108(2), 162-167 (1992-03-01)
Electron microscopic studies of DNA are hampered by difficulties encountered with the spreading of DNA under protein-free conditions. The established and currently popular technique of spreading DNA on carbon membranes treated by glow discharge in an atmosphere of pentylamine is
T Karthick et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 113, 52-66 (2013-05-30)
In this work, the spectroscopic characterization of 2,4,6-tris(dimethylaminomethyl) phenol; a novel promoter factor for DNA has been studied primarily. The FT-IR (4000-400 cm(-1)) and FT-Raman (3500-100 cm(-1)) spectra have been recorded on the solid phase of the title molecule. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service