Skip to Content
Merck
All Photos(1)

Key Documents

QBD10287

Sigma-Aldrich

Amino-dPEG®12-acid

Synonym(s):

Polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H55NO14
Molecular Weight:
617.72
MDL number:
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(O)=O

General description

Amino-dPEG®12-acid is a novel amino acid that can be manipulated, modified, and used like most other amino acids. The single molecular weight dPEG® spacer arms between the terminal amine and the terminal carboxylic acid are of exact length (46.5 Å), allowing for precise spatial control, and they are hydrophilic and non-immunogenic, imparting greater water solubility and hydrodynamic volume to target molecules.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service