Skip to Content
Merck
All Photos(1)

Key Documents

QBD10283

Sigma-Aldrich

Fmoc-N-amido-dPEG®12-acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H65NO16
Molecular Weight:
839.96
MDL number:
UNSPSC Code:
51171641
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reaction type: Pegylations
reagent type: cross-linking reagent

functional group

Fmoc

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

OC(CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCNC(OCC1C2=C(C3=C1C=CC=C3)C=CC=C2)=O)=O

Features and Benefits

Fmoc-N-amido-dPEG®12-acid is a novel Fmoc-protected amino acid containing a single molecular weight PEG (dPEG®) spacer arm. This reagent is used to introduce a hydrophilic, non-immunogenic spacer into a peptide (46.5 Å). Three different lengths of dPEG® allow precise control of spacing and hydrophilicity. Fmoc group can be removed, exposing the terminal amine, using piperidine as with standard peptide synthesis.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service