Skip to Content
Merck
All Photos(1)

Key Documents

QBD10081

Sigma-Aldrich

4-formyl-benzamido-dPEG®12-TFP ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H59F4NO16
Molecular Weight:
897.90
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

>90%

form

solid or viscous liquid

reaction suitability

reactivity: thiol reactive
reagent type: cross-linking reagent
reactivity: amine reactive

polymer architecture

shape: linear
functionality: heterobifunctional

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(C1=CC=C(C=C1)C(NCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCC(OC2=C(C(F)=CC(F)=C2F)F)=O)=O)[H]

InChI

1S/C41H59F4NO16/c42-35-31-36(43)39(45)40(38(35)44)62-37(48)5-7-50-9-11-52-13-15-54-17-19-56-21-23-58-25-27-60-29-30-61-28-26-59-24-22-57-20-18-55-16-14-53-12-10-51-8-6-46-41(49)34-3-1-33(32-47)2-4-34/h1-4,31-32H,5-30H2,(H,46,49)

InChI key

VSRWMAATCHEJKU-UHFFFAOYSA-N

Features and Benefits

4-formyl-benzamido-dPEG®12-TFP ester contains a 4-formyl benzamido group on one end of a 12-unit discrete (single molecular weight) PEG spacer arm and a reactive TFP (2,3,5,6-tetrafluorophenyl) ester group on the other end. TFP ester permits conjugation to a free amine. The 4-formyl-benzamide is used to introduce an aldehyde group into a target molecule. The hydrophilic, non-immunogenic dPEG® spacer provides improved water solubility to the target molecule compared to spacers containing only hydrocarbons.

Legal Information

Products Protected under U.S. Patents # 7,888,536 B2
dPEG is a registered trademark of Quanta BioDesign

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service