Skip to Content
Merck
All Photos(1)

Key Documents

L010002

Sigma-Aldrich

Acetonitrile

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
12191502
PubChem Substance ID:
NACRES:
NA.03
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)

vapor density

1.41 (vs air)

vapor pressure

72.8 mmHg ( 20 °C)

type

for DNA synthesis

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

storage condition

dry at room temperature

impurities

≤30 ppm water content (Karl Fischer)

color

clear

refractive index

n/D 1.342-1.346
n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

solubility

water: soluble (completely)

density

0.786 g/mL at 25 °C (lit.)

format

neat

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Wash solution, containing anhydrous acetonitrile (water content <30 ppm), is used as the overall wash solvent following each step in the synthesis cycle.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Russell J Chedgy et al.
Phytochemistry, 113, 149-159 (2015-01-07)
Salicinoids are phenolic glycosides (PGs) characteristic of the Salicaceae and are known defenses against insect herbivory. Common examples are salicin, salicortin, tremuloidin, and tremulacin, which accumulate to high concentrations in the leaves and bark of willows and poplars. Although their
B Girit et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 71(12), 1859-1868 (2015-06-13)
Bisphenol A (BPA) is an industrial pollutant considered as one of the major endocrine-disrupting chemicals found in natural waters. In the present study, the use of a commercial, air-stable, zero-valent iron (ZVI) powder, consisting of Fe0 surface stabilized nanoparticles was
Takashi Fujii et al.
Nature protocols, 10(9), 1445-1456 (2015-08-28)
Live single-cell mass spectrometry (live MS) provides a mass spectrum that shows thousands of metabolite peaks from a single live plant cell within minutes. By using an optical microscope, a cell is chosen for analysis and a metal-coated nanospray microcapillary
C Nagelreiter et al.
International journal of pharmaceutics, 491(1-2), 162-169 (2015-06-29)
The tape stripping method is a very important tool for dermopharmacokinetic experiments in vitro and the accurate measurement of the removed corneocytes is key for a reliable calculation of a drug's skin penetration behavior. Therefore, various methods to quantify the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service