DWK219902702
Duran® Erlenmeyer Flask
graduated, narrow-mouth neck
Synonym(s):
219902702
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
narrow-mouth neck
feature
graduated
packaging
pkg of 10 ea
manufacturer/tradename
DURAN® 219902702
capacity
150 mL
Looking for similar products? Visit Product Comparison Guide
Application
Media preparation and cultivation, Storing and transport, Mixing several components, Heating Diluting solutions, Measuring liquid amounts, Size 25 ml without Retrace Code und Sizes 150, 200 and 300 ml non-DIN ISO size., Boro 3.3
Features and Benefits
Easy mixing due to conical form ; Easy-to-read scale with graduation marks to estimate the content; Fired-on, highly durable white ceramic
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service