Skip to Content
Merck
All Photos(3)

Key Documents

D107409

Sigma-Aldrich

2′,4′-Dihydroxyacetophenone

99%

Synonym(s):

1-Acetyl-2,4-dihydroxybenzene, 4-Acetylresorcinol, Resacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3COCH3
CAS Number:
Molecular Weight:
152.15
Beilstein:
1282505
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

143-144.5 °C (lit.)

density

1.18 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1ccc(O)cc1O

InChI

1S/C8H8O3/c1-5(9)7-3-2-6(10)4-8(7)11/h2-4,10-11H,1H3

InChI key

SULYEHHGGXARJS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hisao Shimizu et al.
Journal of mass spectrometry : JMS, 47(8), 1015-1022 (2012-08-18)
We investigated the application of a high-resolution Orbitrap mass spectrometer equipped with an electrospray ionization (ESI) source and a matrix-assisted laser desorption/ionization-time-of-flight (MALDI-TOF) mass spectrometer to the metabolite profiling of a model small interfering RNA (siRNA) duplex TSR#34 and compared
M C Almandoz et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 77(1), 51-58 (2010-06-16)
The complexation of Al(III) by 2,4(OH)(2)-acetophenone in methanol and ethanol was investigated using spectroscopic methods (UV-vis) and theoretical procedures (DFT) in order to determine its stoichiometry and stability constant and to analyze the effect of temperature, ionic strength and solvent
Marek Bunse et al.
Chemistry & biodiversity, 17(4), e1900724-e1900724 (2020-02-26)
The investigations reported here focus on an in-depth characterization of the secondary metabolite profile of Sanguisorba officinalis flowers. For this purpose, fresh flowers were extracted with MeOH/H2 O and EtOH/H2 O and the resulting crude extracts fractionated using CH2 Cl2
Katharina Weiss et al.
PloS one, 9(6), e98784-e98784 (2014-06-03)
Effective antimicrobial strategies are essential adaptations of insects to protect themselves, their offspring, and their foods from microbial pathogens and decomposers. Larvae of the emerald cockroach wasp, Ampulex compressa, sanitize their cockroach hosts, Periplaneta americana, with a cocktail of nine
D J Hopper et al.
The Biochemical journal, 344 Pt 2, 397-402 (1999-11-24)
2,4'-Dihydroxyacetophenone dioxygenase (EC 1.13.11.41) was purified to homogeneity from Alcaligenes sp. 4HAP grown on 4-hydroxyacetophenone. Measurements of the M(r) of the native enzyme ranged from 81600 to 87000, whereas values of 21000 and 20379 were given by SDS/PAGE and electrospray

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service