Key Documents
BR41648
BRAND® graduated cylinder low form embossed scale
volume 250 mL, polypropylene
Synonym(s):
Graduated Cylinders, Plastics
About This Item
UNSPSC Code:
41121805
NACRES:
NB.43
Pricing and availability is not currently available.
material
polypropylene
manufacturer/tradename
BRAND 41648
volume
250 mL
General description
The BRAND® Graduated Cylinder, Low Form with Embossed Scale, is designed for precise liquid measurement in laboratory settings. Made from high-clarity polypropylene (PP), this cylinder is calibrated to contain (TC, In), ensuring accurate volume readings. Its low form design enhances stability and ease of use, while the embossed scale provides clear visibility for quick reference. This graduated cylinder can withstand exposure to temperatures up to 80 °C without exceeding permanent error limits, making it suitable for a variety of applications. Reliable and durable, this graduated cylinder is an essential tool for any laboratory.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)