BAH369300050
Sterileware™ sampling spatula
polystyrene, extra long, sterile
All Photos(1)
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
Recommended Products
material
polystyrene
sterility
sterile
packaging
pack of 50 ea
manufacturer/tradename
Bel-Art H36930-0050
Overall L
356 mm (14 in.)
handle L
102 mm (4 in.)
Related Categories
General description
One-piece spatula is molded from rigid, white polystyrene and retains approximately 15cc of material. Shape allows easy transfer of sample to container. Use to sample directly from production line or containers including bags and drums.
- V-shaped spatula is perfect for sampling granulated or semi solid material
- Sterile and individually packaged
- One time use averts cross contamination
- Sharp point will pierce most packaging
Legal Information
Sterileware is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)