BAB610115100
SP Bel-Art® Durac® Bluetooth Thermometer / Hygrometer with 30 day logger
temp. range at -10-50 °C, humidity limit 0-99%, with internal and external sensors
Synonym(s):
Frio-Temp Verification Thermometer / Hygrometer with Internal and External Sensors
About This Item
Recommended Products
material
plastic bottle
stainless steel probe
feature
with internal and external sensors
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B61011-5100
humidity measuring range
10-99%
range of application measuring range
-10-50 °C, resolution: 0.1 °F/C
parameter
temp. range at -10-50 °C
0-99% humidity limit
H × L × W
20 mm × 60 mm × 85 mm
bottle volume
30 mL
probe L
50 mm , 2"
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Monitor critical environments without a physical connection; maximum signal distance is approximately 40 meters
- Record temperature and humidity data for 30 days
- Hourly data recorded for current seven days, then maximum and minimum temperatures and humidity are recorded for next 23 days
- Verification model records accurate temperature reading without sample contamination by using a glass bead filled bottle (included) which mimics the sample
- Alarm sounds to alert user if temperature falls outside of the set parameters
- Data viewable as a table, graph, or calendar; data from up to six data loggers can be plotted on one graph for direct comparison; also downloadable as a CSV file
- Indicators include memory full, active alarm state and low battery; Hygrometer WBGT (Wet Bulb Global Temperature), Heat, Mold and Comfort indices
- App can trigger indicator light for unit identification
- Includes Certificate of Compliance indicating accuracy for 4 temperature and 3 humidity points, and multi-language (English, French, German, Italian, Portuguese and Spanish) instructions
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service