Skip to Content
Merck
All Photos(2)

Key Documents

B68607

Sigma-Aldrich

5-Bromoindole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6BrN
CAS Number:
Molecular Weight:
196.04
Beilstein:
112877
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

powder

mp

90-92 °C (lit.)

SMILES string

Brc1ccc2[nH]ccc2c1

InChI

1S/C8H6BrN/c9-7-1-2-8-6(5-7)3-4-10-8/h1-5,10H

InChI key

VXWVFZFZYXOBTA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui Peng et al.
Environmental science & technology, 50(1), 321-330 (2015-12-01)
Previously unreported natural and synthetic organo-bromine compounds (NSOBCs) have been found to contribute more than 99% of total organic bromine (TOB) in environmental matrices. We recently developed a novel untargeted method (data-independent precursor isolation and characteristic fragment, DIPIC-Frag) and identified
Lisa Cooper et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(23), 6105-6114 (2018-02-03)
Optimisation, scope and mechanism of the platinum-catalysed addition of indoles to indolylallenes is reported here to give 2,3'-BIMs with a novel core structure very relevant for pharmaceutical industry. The reaction is modulated by the electronic properties of the substituents on
Minna K Rahnasto et al.
Bioorganic & medicinal chemistry, 19(23), 7186-7193 (2011-10-25)
The human CYP2A6 enzyme metabolises several xenobiotics including nicotine, the addictive component in tobacco. Reduced activity of CYP2A6, either for genetic reasons or by administering inhibitors of CYP2A6, reduces tobacco smoking. The aim was to design novel inhibitors of CYP2A6
Jeongchan Lee et al.
Nature chemical biology, 17(1), 104-112 (2020-11-04)
Tyrian purple, mainly composed of 6,6'-dibromoindigo (6BrIG), is an ancient dye extracted from sea snails and was recently demonstrated as a biocompatible semiconductor material. However, its synthesis remains limited due to uncharacterized biosynthetic pathways and the difficulty of regiospecific bromination.
Chaitany Jayprakash Raorane et al.
Biomolecules, 10(8) (2020-08-23)
Multi-drug resistant Acinetobacter baumannii is well-known for its rapid acclimatization in hospital environments. The ability of the bacterium to endure desiccation and starvation on dry surfaces for up to a month results in outbreaks of health care-associated infections. Previously, indole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service