Skip to Content
Merck
All Photos(3)

Key Documents

B2403

Sigma-Aldrich

Benz[b]anthracene

98%

Synonym(s):

2,3-Benzanthracene, Naphthacene, Tetracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12
CAS Number:
Molecular Weight:
228.29
Beilstein:
1909299
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

mp

>300 °C (lit.)

Orbital energy

HOMO 5.4 eV 
LUMO 2.7 eV 

OPV Device Performance

ITO/PEDOT:PSS/tetracene/C60/BCP/Al

  • Short-circuit current density (Jsc): 7 mA/cm2
  • Open-circuit voltage (Voc): 0.58 V
  • Fill Factor (FF): 0.57
  • Power Conversion Efficiency (PCE): 2.3 %

semiconductor properties

P-type (mobility=0.4 cm2/V·s)

SMILES string

c1ccc2cc3cc4ccccc4cc3cc2c1

InChI

1S/C18H12/c1-2-6-14-10-18-12-16-8-4-3-7-15(16)11-17(18)9-13(14)5-1/h1-12H

InChI key

IFLREYGFSNHWGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient photovoltaic energy conversion in tetracene-C60 based heterojunctions
Chu, C.; et al.
Applied Physics Letters, 86, 243506-243506 (2005)
Hazuki Morisaki et al.
Nature communications, 5, 5400-5400 (2014-11-08)
Organic crystals are likely to have a large degree of structural relaxation near their surfaces because of the weak inter-molecular interactions. The design of organic field-effect transistors requires a detailed knowledge of the surface relaxation as the carriers usually transfer
Thomas C Allison et al.
The journal of physical chemistry. A, 119(46), 11329-11365 (2015-10-21)
In this article, the first-principles prediction of enthalpies of formation is demonstrated for 669 polycyclic aromatic hydrocarbon (PAH) compounds and a number of related functionalized molecules. It is shown that by extrapolating density functional theory calculations to a large basis
A A Toropov et al.
European journal of medicinal chemistry, 43(4), 714-740 (2007-07-17)
Simplified molecular input line entry system (SMILES) has been utilized in constructing quantitative structure-property relationships (QSPR) for octanol/water partition coefficient of vitamins and organic compounds of different classes by optimal descriptors. Statistical characteristics of the best model (vitamins) are the

Articles

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service