935069
Electrochemical printed electrodes starter kit
pkg of 20
Synonym(s):
BVT-STK
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pkg of 20
General description
Starting kit is the set of twenty different electrochemical sensors to find the best one to fit your application. Starting kit contains 10 types of electrochemical sensors (2 sensors of each type), sensors connector and sensors box with numbered positions and silica gel.
Application
The kit is the set of twenty different electrochemical sensors to find the best one to fit for different applications.
The various application areas of Screen printed electrodes (SPEs) are: (1) for the detection of common environmental pollutants such as metal ions and organic compounds; (2) as enzymatic biosensors, immunosensors and DNA-based sensors for pharmaceutical and clinical usage; (3) also in emerging applications of fabricating lithium-ion batteries, solar cells, and large area LED matrix.
The various application areas of Screen printed electrodes (SPEs) are: (1) for the detection of common environmental pollutants such as metal ions and organic compounds; (2) as enzymatic biosensors, immunosensors and DNA-based sensors for pharmaceutical and clinical usage; (3) also in emerging applications of fabricating lithium-ion batteries, solar cells, and large area LED matrix.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Screen printed silver top electrode for efficient inverted organic solar cells
Materials Research Bulletin, 70, 412-415 (2015)
Applications of screen-printed electrodes in current environmental analysis
Current Opinion in Electrochemistry, 3, 137-143 (2017)
Screen printing of stretchable electrodes for large area LED matrix
Journal of Materials Research, 30, 2271-2278 (2015)
High performance screen-printed electrodes prepared by a green solvent approach for lithium-ion batteries
Journal of Power Sources, 334, 65-77 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service