Skip to Content
Merck
All Photos(1)

Key Documents

93113

Sigma-Aldrich

N-Boc-4,7,10-trioxa-1,13-tridecanediamine

≥95.0% (NT)

Synonym(s):

3-[2-(2-(3-(Boc-amino)propyloxy)ethoxy)ethoxy]propylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H32N2O5
CAS Number:
Molecular Weight:
320.42
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (NT)

form

liquid

reaction suitability

reagent type: cross-linking reagent

density

1.035 g/mL at 20 °C

functional group

Boc
amine

storage temp.

2-8°C

SMILES string

NCCCOCCOCCOCCCNC(OC(C)(C)C)=O

InChI

1S/C15H32N2O5/c1-15(2,3)22-14(18)17-7-5-9-20-11-13-21-12-10-19-8-4-6-16/h4-13,16H2,1-3H3,(H,17,18)

InChI key

WHHYAYNALHPDGJ-UHFFFAOYSA-N

Application

Protected linker.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Songbai Zhang et al.
Pharmaceutics, 12(8) (2020-08-23)
Finding alternatives to gadolinium (Gd)-based contrast agents (CA) with the same or even better paramagnetic properties is crucial to overcome their established toxicity. Herein we describe the synthesis and characterization of entirely organic metal-free paramagnetic macromolecules based on biocompatible oligoethylene
Erkang, F., et al.
Journal of the American Chemical Society, 122, 2663 - 2664 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service