Skip to Content
Merck
All Photos(1)

Key Documents

913383

Sigma-Aldrich

1,3-Dimethylimidazoyl-2-ylidene borane

≥95%

Synonym(s):

(1,3-Dimethyl-1H-imidazol-3-ium-2-yl)trihydroborate, NHC-borane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11BN2
CAS Number:
Molecular Weight:
109.97
MDL number:

Assay

≥95%

form

crystals

availability

available only in USA

mp

138-139 °C

storage temp.

2-8°C

SMILES string

[B-H]c1[n+](cc[n]1C)C

InChI key

WDITXRGBBBKEET-UHFFFAOYSA-N

Application

1,3-Dimethylimidazoyl-2-ylidene borane is stable NHC-borane developed in the Curran lab. It is a diverse reductant as well as a versatile hydroboration reagent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Dai et al.
Journal of the American Chemical Society, 141(31), 12355-12361 (2019-07-12)
Reactions of readily available alkenyl triflates with N-heterocyclic carbene (NHC)-boranes in the presence of diisopropyl ethyl amine provided about three dozen stable α-NHC-boryl ketones. Isolated yields were typically 40-56% for B-unsubstituted NHC-boranes (NHC-BH3), and somewhat lower for NHC-boranes with B-substituents
Daniel Subervie et al.
Angewandte Chemie (International ed. in English), 57(32), 10251-10256 (2018-06-22)
1,3-Dimethylimidazol-2-ylidene difluoroborane (NHC-BF2 H) was prepared in a one-pot, two-step reaction from the parent ligated borane (NHC-BH3 ). The derived difluoroboryl radical (NHC-BF2. ) was generated by laser flash photolysis experiments and characterized by UV spectroscopy and rate-constant measurements. It is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service