Skip to Content
Merck
All Photos(4)

Key Documents

901906

Sigma-Aldrich

SPhos

95%

Synonym(s):

2-Dicyclohexylphosphino-2′,6′-dimethoxybiphenyl, S Phos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H35O2P
CAS Number:
Molecular Weight:
410.53
MDL number:
UNSPSC Code:
12161600
NACRES:
NA.22

Assay

95%

form

powder or crystals

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

reagent type: ligand

mp

164-166 °C (lit.)
165.5 °C

functional group

phosphine

SMILES string

COc1cccc(OC)c1-c2ccccc2P(C3CCCCC3)C4CCCCC4

InChI

1S/C26H35O2P/c1-27-23-17-11-18-24(28-2)26(23)22-16-9-10-19-25(22)29(20-12-5-3-6-13-20)21-14-7-4-8-15-21/h9-11,16-21H,3-8,12-15H2,1-2H3

InChI key

VNFWTIYUKDMAOP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SPhos [2-Dicyclohexylphosphino-2′, 6′-dimethoxybiphenyl] is an air-stable, electron-rich biaryl phosphine ligand developed by the Buchwald group to enhance the reactivity of palladium catalysis during cross-coupling reactions.

Application

Highly universal ligand for Suzuki-Miyaura coupling; aryl chlorides, hindered biaryls, heterobiaryls.
SPhos may be used as a ligand in the following processes:
  • Palladium catalyzed Suzuki-Miyaura cross-coupling reaction between Boc-protected aminomethyltrifluoroborate and aryl chlorides or hetaryl chlorides to form the corresponding aminomethylarenes.
  • Palladium catalyzed Suzuki-Miyaura cross-coupling reaction between 4-methyl-substituted piperidinylzinc reagent and different aryl or heteroaryl iodides to form various substituted piperidines.
  • Intramolecular Suzuki-Miyaura coupling to form the 18-membered macrocyclic ring during the multi-step synthesis of riccardin C.
Utilized in conjunction with palladium to form a highly active catalyst for C-N bond formation

Legal Information

Usage subject to Patents: US 6307087; EP 1097158; JP 5758844; CA 2336691

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd-catalyzed Suzuki?Miyaura reactions of aryl halides using bulky biarylmonophosphine ligands
Altman, Ryan A., and Stephen L. Buchwald
Nature Protocols, 2.12 (2007)
Evidence for the formation and structure of palladacycles during Pd-catalyzed C-N bond formation with catalysts derived from bulky monophosphinobiaryl ligands.
Eric R Strieter et al.
Angewandte Chemie (International ed. in English), 45(6), 925-928 (2005-12-29)
Synthesis and Suzuki?Miyaura Cross-Coupling Reactions of Potassium Boc-Protected Aminomethyltrifluoroborate with Aryl and Hetaryl Halides
Molander, Gary A., and Inji Shin.
Organic Letters, 13.15, 3956-3959 (2011)
Synthesis of riccardin C and its seven analogues. Part 1: The role of their phenolic hydroxy groups as LXR? agonists
Hioki, Hideaki, et al.
Bioorganic & Medicinal Chemistry Letters, 19.3, 738-741 (2009)
Synthesis and Suzuki-Miyaura Cross-Coupling Reactions of Potassium Boc-Protected Aminomethyltrifluoroborate with Aryl and Hetaryl Halides.
Molander GA and Shin I.
Organic Letters, 13(15), 3956-3959 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service