Skip to Content
Merck
All Photos(1)

Key Documents

861634

Sigma-Aldrich

2,4-Diamino-6-(hydroxymethyl)pteridine

95%

Synonym(s):

2,4-Diamino-6-pteridinemethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N6O
CAS Number:
Molecular Weight:
192.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

SMILES string

Nc1nc(N)c2nc(CO)cnc2n1

InChI

1S/C7H8N6O/c8-5-4-6(13-7(9)12-5)10-1-3(2-14)11-4/h1,14H,2H2,(H4,8,9,10,12,13)

InChI key

CYNARAWTVHQHDI-UHFFFAOYSA-N

Gene Information

rat ... Dhfr(24312)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ming-Hao Hsu et al.
Journal of hazardous materials, 373, 468-475 (2019-04-03)
Methotrexate, a chemotherapeutic agent, was found to undergo self-sensitized photodegradation in aqueous environments. As the initial concentration increased, methotrexate was able to enhance its own direct photolysis reaction not only in DI but also in natural waters. The methotrexate degradation
Webber Wei-Po Lai et al.
Water research, 112, 157-166 (2017-02-06)
Bicarbonate anion (HCO

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service