Skip to Content
Merck
All Photos(1)

Key Documents

76840

Sigma-Aldrich

Pentamethyldisiloxane

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H16OSi2
CAS Number:
Molecular Weight:
148.35
Beilstein:
1734568
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0%

form

liquid

refractive index

n20/D 1.373

bp

86 °C (lit.)

density

0.760 g/mL at 20 °C (lit.)

SMILES string

[H][Si](C)(C)O[Si](C)(C)C

InChI

1S/C5H16OSi2/c1-7(2)6-8(3,4)5/h7H,1-5H3

InChI key

AOJHDNSYXUZCCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sijia Zheng et al.
Macromolecular rapid communications, 42(5), e2000375-e2000375 (2020-08-15)
Most silicone elastomers are thermosets. As a response to the new paradigm of polymer recyclability, the development of silicone elastomers that can be reversibly and repeatedly cured and uncrosslinked using redox conditions is reported. Thiopropyl-modified silicones are oxidized to elastomers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service