Skip to Content
Merck
All Photos(1)

Key Documents

750832

Sigma-Aldrich

2-Keto-4-methylpentanoic acid-1-13C

99 atom % 13C, 98% (CP)

Synonym(s):

2-Ketoisocaproic acid-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CO13CO2H
Molecular Weight:
131.13
UNSPSC Code:
12352115
PubChem Substance ID:

isotopic purity

99 atom % 13C

Assay

98% (CP)

form

liquid

mass shift

M+1

storage temp.

−20°C

SMILES string

CC(C)CC(=O)[13C](O)=O

InChI

1S/C6H10O3/c1-4(2)3-5(7)6(8)9/h4H,3H2,1-2H3,(H,8,9)/i6+1

InChI key

BKAJNAXTPSGJCU-PTQBSOBMSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Russell E Ericksen et al.
Cell metabolism, 29(5), 1151-1165 (2019-01-22)
Tumors display profound changes in cellular metabolism, yet how these changes aid the development and growth of tumors is not fully understood. Here we use a multi-omic approach to examine liver carcinogenesis and regeneration, and find that progressive loss of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service