Skip to Content
Merck
All Photos(2)

Key Documents

747157

Sigma-Aldrich

Potassium pentafluroroethyltrifluoroborate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2BF8K
CAS Number:
Molecular Weight:
225.92
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

252-257 °C

functional group

fluoro

storage temp.

2-8°C

SMILES string

F[B-](F)(F)C(F)(F)C(F)(F)F.[K+]

InChI

1S/C2BF8.K/c4-1(5,2(6,7)8)3(9,10)11;/q-1;+1

InChI key

PSJPJAFBTMLFFX-UHFFFAOYSA-N

Application

Organotrifluoroborate involved in:
  • Suzuki Miyaura cross-coupling reactions, and polymerization reactions
  • Synthesis of photonic crystals
  • Synthesis of sensitizers for dye-sensitized solar cells
  • Mannich / diastereoselective hydroamination reaction sequence

Organotrifluoroborates as versatile and stable boronic acid surrogates

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jenny M Baxter Vu et al.
Organic letters, 13(15), 4056-4059 (2011-07-14)
A new two-step synthesis of highly substituted pyrrolidines has been developed. Chiral silane Lewis acid promoted enantioselective Mannich reactions of silyl ketene imines with acylhydrazones may be used to access bishomoallylic benzoic hydrazides that in turn may be cyclized to
R. Grisorio;
Journal of Polymer Science: Part A, General Papers, 49, 842-847 (2011)
Roberto Grisorio et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 16(27), 8054-8061 (2010-06-04)
The mechanism of the Suzuki-Heck (SuHe) polymerisation of 2,7-dibromo-9,9-di(n-dodecyl)fluorene (1) with potassium vinyl trifluoroborate (PVTB) for the synthesis of poly(fluorenylene vinylene)s (PFVs) has been investigated. In the first stage, a palladium-catalysed chain-growth AA/B(C)-type polycondensation occurs, as evidenced by the linear
Emilio Alacid et al.
The Journal of organic chemistry, 74(6), 2321-2327 (2009-02-17)
Potassium vinyl and alkenyltrifluoroborates are cross-coupled with aryl and heteroaryl bromides using 1 mol % Pd loading of 4-hydroxyacetophenone oxime derived palladacycle or Pd(OAc)2 as precatalysts, K2CO3 as base, and TBAB as additive and water reflux under conventional or microwave
S. Achelle;
Journal of Polymer Science: Part A, General Papers, 48, 2659-2665 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service