Skip to Content
Merck
All Photos(1)

Key Documents

742937

Sigma-Aldrich

1,4-Diazabicyclo[2.2.2]octane bis(sulfur dioxide) adduct

≥95% (sulfur, elemental analysis)

Synonym(s):

DABCO•(SO2)2, DABSO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O4S2
CAS Number:
Molecular Weight:
240.30
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (sulfur, elemental analysis)

form

powder

composition

active SO2, ~50%
carbon, 28.3-31.6%

storage temp.

2-8°C

SMILES string

[O-]S(=O)[N+]12CC[N+](CC1)(CC2)S([O-])=O

InChI

1S/C6H12N2O4S2/c9-13(10)7-1-2-8(5-3-7,6-4-7)14(11)12/h1-6H2

InChI key

RWISEVUOFYXWFO-UHFFFAOYSA-N

Application

1,4-Diazabicyclo[2.2.2]octane bis(sulfur dioxide) (DABSO) is a charge transfer complex that can be used as a sulfur dioxide surrogate:
  • In palladium-catalyzed aminosulfonylation process.
  • In reaction with aryl bromides to synthesize sodium aryl sulfinates.

It can also be used to activate DMSO and o-vinylanilines for the synthesis of N-aryl-1H-benzo[d]imidazol-1-amine and 4-aryl quinolines, respectively.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Palladium-catalyzed annulation of 2-(aryldiazenyl) aniline with dimethyl sulfoxide to access N-aryl-1H-benzo [d] imidazol-1-amine.
Wang H, et al.
Tetrahedron Letters, 58(40), 3875-3878 (2017)
Palladium-catalyzed aminosulfonylation of aryl halides.
Nguyen B, et al.
Journal of the American Chemical Society, 132(46), 16372-16373 (2010)
Bao Nguyen et al.
Journal of the American Chemical Society, 132(46), 16372-16373 (2010-10-30)
The palladium-catalyzed three-component coupling of aryl iodides, sulfur dioxide, and hydrazines to deliver aryl N-aminosulfonamides is described. The colorless crystalline solid DABCO·(SO(2))(2) was used as a convenient source of sulfur dioxide. The reaction tolerates significant variation of both the aryl
Synthesis of sodium aryl sulfinates from aryl bromides employing 1, 4-diazabicyclo [2.2. 2] octane bis (sulfur dioxide) adduct (DABSO) as a bench-stable, gas-free alternative to SO2.
Skillinghaug B, et al.
Tetrahedron Letters, 57(5), 533-536 (2016)
Carbon annulation of ortho-vinylanilines with dimethyl sulfoxide to access 4-aryl quinolines.
Yuan J, et al.
Organic & Biomolecular Chemistry, 15(6), 1334-1337 (2017)

Related Content

Research in the Willis group is focused on the development of new catalysts and reactions for synthetic chemistry. The group is particularly interested in addressing synthetic challenges that are applicable to the pharmaceutical and agrochemical industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service