Skip to Content
Merck
All Photos(1)

Key Documents

741868

Sigma-Aldrich

Buchwald Palladacycle Precatalyst Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161600
NACRES:
NA.22
Pricing and availability is not currently available.

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

Kit Components Also Available Separately

Product No.
Description
SDS

  • 704954XPhos Pd G1 250 mgSDS

  • 707589RuPhos Pd G1 Methyl -Butyl Ether Adduct, 95% 250 mgSDS

  • 718750BrettPhos Pd G1, Methyl t-Butyl Ether Adduct, may contain up to 1 mole equivalent of MTBE, 97% 100 mgSDS

  • 704946SPhos Pd G1, Methyl t-Butyl Ether Adduct 250 mgSDS

  • 708739tBuXPhos Pd G1 250 mgSDS

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service