725676
Poly(ethylene glycol) diacrylamide
average MN 3,700, acrylamide, ≤1,500 ppm HQ as inhibitor
Synonym(s):
Polyethylene glycol
About This Item
Recommended Products
Product Name
Poly(ethylene glycol) diacrylamide, average Mn 3,700, contains ≤1,500 ppm HQ as inhibitor
form
solid
mol wt
average Mn 3,700
contains
≤1,500 ppm HQ as inhibitor
transition temp
Tm 50-54 °C
Ω-end
acrylamide
α-end
acrylamide
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Preparation Note
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.
Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.
Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.
Scaffold patterning with poly(ethylene glycol)-based hydrogels for cell presence in 2D and 3D environments on photoactive substrates.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service