Skip to Content
Merck
All Photos(1)

Key Documents

706035

Sigma-Aldrich

Aldosterone-2,2,4,6,6,21,21-d7

≥98 atom % D (based on d7), ≥98% (CP)

Synonym(s):

11β,21-Dihydroxy-3,20-dioxo-4-pregnen-18-al-D7, 11β,21-Dihydroxypregn-4-ene-3,18,20-trione-D7, 11β,21-Dihyroxy-3,20-dioxo-4-pregnen-18-al-d7 (2,2,4,6,6,21,21-d7), 11β,21-Dihyroxypregn-4-ene-3,18,20-trione-d7 (2,2,4,6,6,21,21-d7), 18-Aldocorticosterone-2,2,4,6,6,21,21-d7, Deuterated aldocorticosterone, Deuterated aldosterone, Reichstein X (2,2,4,6,6,21,21-d7), Reichstein X-D7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21D7H21O5
CAS Number:
Molecular Weight:
367.49
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.24

isotopic purity

≥98 atom % D (based on d7)

Assay

≥98% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

storage temp.

−20°C

SMILES string

[2H]C1=C2C([2H])([2H])C[C@H]3[C@@H]4CCC([2H])(C(=O)C(O)([*])[*])[C@]4(C[C@H](O)[C@@H]3[C@@]2(C)CC([2H])([2H])C1=O)C=O

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liyun Zhang et al.
Frontiers in pharmacology, 12, 646240-646240 (2021-06-29)
Objective: Accumulating evidence suggested that resveratrol (RES) could protect against adverse cardiac remodeling induced by several cardiovascular diseases. However, the role of RES in the setting of heart failure with preserved ejection fraction (HFpEF) and the underlying mechanisms of its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service